Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE4C Antibody, Novus Biologicals™
SDP

Catalog No. NB018171 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018171 0.025 mg
NB300681 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018171 Supplier Novus Biologicals Supplier No. NB3006810.025MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

PDE4C Polyclonal specifically detects PDE4C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen PDE4C
Applications Immunoprecipitation, Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunoprecipitation 1:20
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias cAMP-specific 3'-5'-cyclic phosphodiesterase 4C, DPDE1dunce, Drosophila), EC 3.1.4, EC 3.1.4.17, MGC126222, PDE21, phosphodiesterase 4C, cAMP-specific, phosphodiesterase 4C, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E1), phosphodiesterase 4C, cAMP-specific (phosphodiesterase E1 dunce homolog, phosphodiesterase E1 dunce homolog
Gene Symbols PDE4C
Host Species Rabbit
Immunogen Synthetic cyclic peptide (carboxy-terminal peptide common to all PDE4C proteins).
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5143
Test Specificity This detects 3 isoforms of PDE4C subclasses (PDE4C1, PDE4C2 and PDE4C3).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.