Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE5A Antibody, Novus Biologicals™
SDP

Catalog No. NB018164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018164 0.025 mg
NB300655005 0.05 mg
NB300655 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018164 Supplier Novus Biologicals Supplier No. NB3006550.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE5A Polyclonal specifically detects PDE5A in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen PDE5A
Applications Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:250
Formulation Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Gene Alias CGB-PDE, cGMP-binding cGMP-specific 3'-5'-cyclic nucleotide phosphodiesterase, cGMP-binding cGMP-specific phosphodiesterase, cGMP-specific 3'-5'-cyclic phosphodiesterase, cGMP-specific phosphodiesterase type 5A, CN5A, EC 3.1.4, EC 3.1.4.35, PDE5, phosphodiesterase 5A, cGMP-specific, phosphodiesterase isozyme 5
Gene Symbols PDE5A
Host Species Rabbit
Immunogen Synthetic cyclic peptide (common to all PDE5A variants) (Rat).
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8654
Test Specificity This detects 99 and 89 kDa PDE5A proteins in various tissues.
Target Species Human, Rat, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.