Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE5A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$268.00 - $706.50
Specifications
Antigen | PDE5A |
---|---|
Dilution | Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:250 |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018164
![]() |
Novus Biologicals
NB3006550.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NB300655005
![]() |
Novus Biologicals
NB3006550.05MG |
0.05 mg |
Each for $426.00
|
|
|||||
NB300655
![]() |
Novus Biologicals
NB300655 |
0.1 mg |
Each for $706.50
|
|
|||||
Description
PDE5A Polyclonal specifically detects PDE5A in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
PDE5A | |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
8654 | |
Synthetic cyclic peptide (common to all PDE5A variants) (Rat). | |
Primary |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:250 | |
Polyclonal | |
Rabbit | |
PDE5A | |
IgG | |
This detects 99 and 89 kDa PDE5A proteins in various tissues. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title