Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE6 beta Antibody, Novus Biologicals™
SDP

Catalog No. NB1205663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205663 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205663 Supplier Novus Biologicals Supplier No. NB1205663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE6 beta Polyclonal antibody specifically detects PDE6 beta in Human, Mouse, Bovine, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen PDE6 beta
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Formulation PBS containing 1mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. P23440
Gene Alias CSNB3, EC 3.1.4, EC 3.1.4.35, GMP-PDE beta, PDEBrod cGMP-phosphodiesterase beta-subunit, phosphodiesterase 6B, cGMP-specific, rod, beta, rd1, rod cGMP-specific 3'-5'-cyclic phosphodiesterase subunit beta, RP40CSNBAD2
Gene Symbols PDE6B
Host Species Rabbit
Immunogen Synthetic Peptides: H(20) Q Y F G (K/R) K L S P E N V A G A C(36)
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5158
Test Specificity Detects PDE6B from mouse retinal extracts.
Target Species Human, Mouse, Bovine, Canine, Monkey
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.