Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE6A Antibody, Novus Biologicals™
SDP

Catalog No. NB1205660 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205660 Supplier Novus Biologicals Supplier No. NB1205660

Rabbit Polyclonal Antibody

PDE6A Polyclonal specifically detects PDE6A in Bovine, Sheep samples. It is validated for Western Blot.

Specifications

Antigen PDE6A
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS with 0.05% Sodium Azide
Gene Alias CGPR-A, EC 3.1.4, EC 3.1.4.35, GMP-PDE alpha, PDE V-B1, PDEAcGMP phosphodiesterase alpha subunit, phosphodiesterase 6A, cGMP-specific, rod, alpha, rod cGMP-specific 3'-5'-cyclic phosphodiesterase subunit alpha, rod photoreceptor cGMP phosphodiesterase alpha subunit, RP43
Gene Symbols PDE6A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(16) P Q F A K E Y F D R K L R(29) C of human PDE6 alpha.
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5145
Test Specificity PDE6 alphaÆ
Target Species Bovine, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.