Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE6G Antibody (OTI2F5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273298IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273298IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273298IR Supplier Novus Biologicals Supplier No. NBP273298IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE6G Monoclonal specifically detects PDE6G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PDE6G
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2F5
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686C0587, GMP-PDE gamma, MGC125749, PDEG, phosphodiesterase 6G, cGMP-specific, rod, gamma, retinal rod rhodopsin-sensitive cGMP 3′-5′-cyclic phosphodiesterase subunit gamma, rod cG-PDE G, RP57
Gene Symbols PDE6G
Host Species Mouse
Immunogen Full length recombinant protein of human PDE6G (NP_002593) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5148.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.