Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE6H Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | PDE6H |
---|---|
Dilution | Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Immunohistochemistry-Frozen -Reported in scientific literature (PMID:35024589) |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PDE6H Polyclonal antibody specifically detects PDE6H in Human, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
PDE6H | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Signal Transduction, Vision | |
PBS (pH 7.2) and 40% Glycerol | |
5149 | |
IgG | |
Protein A purified |
Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000, Immunohistochemistry-Frozen -Reported in scientific literature (PMID:35024589) | |
Polyclonal | |
Purified | |
RUO | |
Human, Monkey | |
EC 3.1.4.17, EC 3.1.4.35, GMP-PDE gamma, phosphodiesterase 6H, cGMP-specific, cone, gamma, RCD3, retinal cone rhodopsin-sensitive cGMP 3'-5'-cyclic phosphodiesterase subunitgamma | |
This antibody was developed against a recombinant protein corresponding to amino acids: PRKGPPKFKQRQTRQFKSKPPKKGVKGFGDDIPGMEGLGTDITVICPWEAFSHLELHELA | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title