Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE7A Antibody, Novus Biologicals™
SDP

Catalog No. NB300639005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300639005 0.05 mg
NB018159 0.025 mg
NB300639 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB300639005 Supplier Novus Biologicals Supplier No. NB3006390.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE7A Polyclonal specifically detects PDE7A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PDE7A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunoprecipitation 1:250
Formulation Tris/Glycine buffer, pH 7.4; cryoprotective agents; HEPES; stabilizing protein (BSA 0.5%), and glycerol (30%). with 0.02% sodium azide
Gene Alias EC 3.1.4, EC 3.1.4.17, HCP1high affinity cAMP-specific 3′-5′-cyclic phosphodiesterase 7A, PDE7, phosphodiesterase 7A, phosphodiesterase isozyme 7, TM22
Gene Symbols PDE7A
Host Species Rabbit
Immunogen Synthetic cyclic peptide (common to all PDE7A variants).
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 5150
Test Specificity PDE7A
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.