Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE8A Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291203 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291203 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291203 Supplier Novus Biologicals Supplier No. NBP152651APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE8A Polyclonal antibody specifically detects PDE8A in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PDE8A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias cAMP-specific cyclic nucleotide phosphodiesterase 8A, EC 3.1.4.17, FLJ16150, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, high-affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, HsT19550, phosphodiesterase 8A
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 779 and 829 of human Phosphodiesterase 8A using the numbering given in entry NP_002596.1 (GeneID 5151).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5151
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.