Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE8A Antibody (EM-52), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB002564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002564 Supplier Novus Biologicals Supplier No. NBP262213AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDE8A Monoclonal specifically detects PDE8A in Human samples. It is validated for Western Blot.

Specifications

Antigen PDE8A
Applications Western Blot
Classification Monoclonal
Clone EM-52
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cAMP-specific cyclic nucleotide phosphodiesterase 8A, EC 3.1.4.17, FLJ16150, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, high-affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8A, HsT19550, phosphodiesterase 8A
Gene Symbols PDE8A
Host Species Mouse
Immunogen Human PDE8a
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5151
Test Specificity The antibody MEM-115 recognizes an epitope in the D1 domain of CD4 antigen, a 55 kDa transmebrane glycoprotein expressed on a subset of T lymphocytes and also on monocytes, tissue macrophages and granulocytes. It is negative in Western blotting even with non-reduced samples of cell lysates.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.