Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE8B Antibody, Novus Biologicals™
SDP

Catalog No. NB300644005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300644005 0.05 mg
NB018161 0.025 mg
NB300644 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB300644005 Supplier Novus Biologicals Supplier No. NB3006440.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE8B Polyclonal specifically detects PDE8B in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen PDE8B
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 0.50 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:10000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:10-1:500
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Alias 3′-5′ cyclic nucleotide phosphodiesterase 8B, ADSD, Cell proliferation-inducing gene 22 protein, EC 3.1.4.17, FLJ11212, high affinity cAMP-specific and IBMX-insensitive 3′-5′-cyclic phosphodiesterase8B, hsPDE8B, phosphodiesterase 8B, PPNAD3
Gene Symbols PDE8B
Host Species Rabbit
Immunogen Synthetic cyclic peptide common to all PDE8B variants.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 8622
Test Specificity This has no cross-reactivity against PDE8A protein or any other PDE family members.
Target Species Human, Mouse, Rat, Bovine, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.