Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDE8B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PDE8B |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PDE8B Polyclonal specifically detects PDE8B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PDE8B | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism, Signal Transduction | |
3'-5' cyclic nucleotide phosphodiesterase 8B, ADSD, Cell proliferation-inducing gene 22 protein, EC 3.1.4.17, FLJ11212, high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8B, hsPDE8B, phosphodiesterase 8B, PPNAD3 | |
PDE8B | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
8622 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RRHCCSSAEAETQTCYTSVKQVSSAEVRIGPMRLTQDPIQVLLIFAKEDSQSDGFWWACDRAGYRCNIARTPESALECFLD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title