Learn More
Description
Specifications
Specifications
| Antigen | PDE9A |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, EC 3.1.4.1710phosphodiesterase PDE9A21, EC 3.1.4.35, FLJ90181, high affinity cGMP-specific 3'-5'-cyclic phosphodiesterase 9A, HSPDE9A2, phosphodiesterase 9A |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant PDE9A (Pro181-Lys506). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
