Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDE9A Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB323344 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323344 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323344 Supplier Novus Biologicals Supplier No. NBP299870MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDE9A Polyclonal antibody specifically detects PDE9A in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen PDE9A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, CGMP-specific 3'-5'-cyclic phosphodiesterase type 9, EC 3.1.4.1710phosphodiesterase PDE9A21, EC 3.1.4.35, FLJ90181, high affinity cGMP-specific 3'-5'-cyclic phosphodiesterase 9A, HSPDE9A2, phosphodiesterase 9A
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human PDE9A (Accession#: O76083-2; Pro181-Lys506)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5152
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.