Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-B Antibody (MM0014-5F66) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB017983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017983 0.025 mg
NB11060980 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017983 Supplier Novus Biologicals Supplier No. NB110609800.025MG

Mouse Monoclonal Antibody

PDGF-B Monoclonal specifically detects PDGF-B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen PDGF-B
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0014-5F66
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. P01127
Gene Alias B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV
Gene Symbols PDGFB
Host Species Mouse
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human PDGF BB recombinant protein.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5155
Test Specificity This antibody shows approximately some degree (40%) cross-reactivity with rPDGF-AB and no cross-reactivity with recombinant PDGF-AA.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.