Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-B Antibody (PDGFB/3072), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB390337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390337 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390337 Supplier Novus Biologicals Supplier No. NBP308823JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDGF-B Monoclonal antibody specifically detects PDGF-B in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen PDGF-B
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone PDGFB/3072
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV
Host Species Mouse
Immunogen Recombinant fragment of human PDGF-B protein (around aa 27-158) (exact sequence is proprietary) (Uniprot: P01127)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5155
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.