Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-C Antibody (004), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB088599 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088599 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088599 Supplier Novus Biologicals Supplier No. NBP289373DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PDGF-C Monoclonal specifically detects PDGF-C in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDGF-C
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 4
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human PDGF-C (NP_057289.1; Val235-Gly345).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 56034
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.