Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-C Antibody (5F12), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB360874 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360874 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360874 Supplier Novus Biologicals Supplier No. NBP221956MFV450

Mouse Monoclonal Antibody

PDGF-C Monoclonal antibody specifically detects PDGF-C in Human samples. It is validated for Western Blot, Neutralization

Specifications

Antigen PDGF-C
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 5F12
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias FALLOTEIN, PDGF-C, platelet derived growth factor C, platelet-derived growth factor C, SCDGFSpinal cord-derived growth factor, secretory growth factor-like protein, VEGF-E
Host Species Mouse
Immunogen Human recombinant PDGF-C
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 56034
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.