Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF-D Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC1451100UG
Description
Anti-PDGF-D is a rabbit polyclonal antibody that detects Platelet-derived growth factor D in human cells and has been tested for use in Western Blotting.
Specifications
PDGF-D | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This rabbit polyclonal antibody detects Platelet-derived growth factor D in human cells. It targets an epitope within the growth factor domain from the C-terminal half. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG |
Western Blot | |
Please refer to lot specific datasheet. | |
PDGFD,IEGF,SCDGFB,MSTP036 | |
A linear peptide corresponding to 18 amino acids from the C-terminal half of human Platelet-derived growth factor D. | |
100 μg | |
Apoptosis and Cancer | |
NP_079484 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction