Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF-D Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.34 - $469.82
Specifications
Antigen | PDGF-D |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABC1451-25UG
|
MilliporeSigma
ABC145125UG |
25 μg |
Each for $192.34
|
|
|||||
ABC1451100
|
MilliporeSigma
ABC1451100UG |
100 μg |
Each for $469.82
|
|
|||||
Description
Anti-PDGF-D is a rabbit polyclonal antibody that detects Platelet-derived growth factor D in human cells and has been tested for use in Western Blotting.Specifications
PDGF-D | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis and Cancer | |
NP_079484 | |
A linear peptide corresponding to 18 amino acids from the C-terminal half of human Platelet-derived growth factor D. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Human | |
PDGFD,IEGF,SCDGFB,MSTP036 | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody detects Platelet-derived growth factor D in human cells. It targets an epitope within the growth factor domain from the C-terminal half. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title