Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-D/SCDGFB Antibody (OTI1C1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27330A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27330A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27330A32 Supplier Novus Biologicals Supplier No. NBP273305AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDGF-D/SCDGFB Monoclonal specifically detects PDGF-D/SCDGFB in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen PDGF-D/SCDGFB
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1C1
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias IEGFMSTP036, Iris-expressed growth factor, PDGF-D, platelet derived growth factor D, SCDGF-BMGC26867, SCDGFBplatelet-derived growth factor D, spinal cord derived growth factor B, Spinal cord-derived growth factor B, spinal cord-derived growth factor-B
Gene Symbols PDGFD
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 19-247 of human PDGF-D/SCDGFB (NP_079484) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 80310.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.