Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF-D/SCDGFB Antibody (OTI1C1) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP273305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273305 Supplier Novus Biologicals Supplier No. NBP273305
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDGF-D/SCDGFB Monoclonal specifically detects PDGF-D/SCDGFB in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen PDGF-D/SCDGFB
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone OTI1C1
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:100, CyTOF-ready
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias IEGFMSTP036, Iris-expressed growth factor, PDGF-D, platelet derived growth factor D, SCDGF-BMGC26867, SCDGFBplatelet-derived growth factor D, spinal cord derived growth factor B, Spinal cord-derived growth factor B, spinal cord-derived growth factor-B
Gene Symbols PDGFD
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 19-247 of human PDGFD(NP_079484) produced in E.coli.
Molecular Weight of Antigen 40.2 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 80310
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.