Learn More
Description
Specifications
Specifications
| Antigen | PDGF R beta |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 18A2 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Gene Alias | beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide |
| Gene Symbols | PDGFRB |
| Host Species | Mouse |
| Immunogen | CD140b-transfected NIH 3T3 cells |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
