Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF R beta Antibody (18A2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP47232646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP47232646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP47232646 Supplier Novus Biologicals Supplier No. NBP147232JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDGF R beta Monoclonal specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDGFRbeta
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 18A2
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Gene Symbols PDGFRB
Host Species Mouse
Immunogen CD140b-transfected NIH 3T3 cells
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Oncogenes, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5159
Test Specificity The monoclonal antibody 18A2 recognizes CD140b PDGF-RB, the 180-190 kDa beta chain of platelet-derived growth factor receptor, which is widely expressed on a variety of mesenchymal-derived cells and plays pro-proliferative roles.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.