Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF R beta Antibody (APB5), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NBP43349646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP43349646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP43349646 Supplier Novus Biologicals Supplier No. NBP143349JF646
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen PDGFRbeta
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone APB5
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Gene Symbols PDGFRB
Host Species Rat
Immunogen The immunogen for this antibody was PDGF receptor beta.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Oncogenes, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5159
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.