Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF R beta Antibody (PR7212), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB175082 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175082 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175082 Supplier Novus Biologicals Supplier No. FAB1263L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDGF R beta Monoclonal specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen PDGF R beta
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PR7212
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Host Species Mouse
Immunogen Human skin fibroblast membrane extracts
Purification Method Protein A or G purified from ascites
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Oncogenes, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5159
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.