Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGF R beta Antibody (PR7212), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB314657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314657 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314657 Supplier Novus Biologicals Supplier No. FAB1263PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDGF R beta Monoclonal specifically detects PDGF R beta in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PDGF R beta
Applications Flow Cytometry
Classification Monoclonal
Clone PR7212
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide
Host Species Mouse
Immunogen Human skin fibroblast membrane extracts
Purification Method Protein A or G purified from ascites
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Oncogenes, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5159
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.