Learn More
Description
Specifications
Specifications
| Antigen | PDGF R beta |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 1% BSA, 20% Glycerol (pH7) |
| Gene Alias | beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide |
| Host Species | Rabbit |
| Immunogen | Carrier-protein conjugated synthetic peptide corresponding to residues around human PDGF R beta (phospho Tyr857). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
