Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFR alpha Antibody (189208), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB174831 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174831 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174831 Supplier Novus Biologicals Supplier No. FAB1062Z
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

PDGFR alpha Monoclonal specifically detects PDGFR alpha in Mouse samples. It is validated for Western Blot.

Specifications

Antigen PDGFR alpha
Applications Western Blot
Classification Monoclonal
Clone 189208
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse PDGF R alpha , Leu25-Glu524 (Asp65Glu, Gly439Ala, Thr440Ala), predicted, Accession # P26618
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5156
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.