Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFR alpha Antibody (BLR128H), Novus Biologicals™
SDP

Catalog No. NB188051 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188051 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB188051 Supplier Novus Biologicals Supplier No. NBP314711
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation

Specifications

Antigen PDGFR alpha
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone BLR128H
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation Borate Buffered Saline (BBS) pH 8.2 with, BSA Free with 0.09% Sodium Azide
Gene Alias alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Host Species Rabbit
Immunogen residues 24-525 (ECD)
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5156
Target Species Human, Mouse
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.