Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFR alpha Antibody (PRa292), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB175092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175092 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175092 Supplier Novus Biologicals Supplier No. FAB1264L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PDGFR alpha Monoclonal antibody specifically detects PDGFR alpha in Human samples. It is validated for Flow Cytometry

Specifications

Antigen PDGFR alpha
Applications Flow Cytometry
Classification Monoclonal
Clone PRa292
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Host Species Mouse
Immunogen Human osteosarcoma cell membrane extracts
Purification Method Protein A or G purified from ascites
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5156
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.