Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFR alpha Antibody (RM0004-3G28) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB11061020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11061020 0.1 mg
NB017999 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11061020 Supplier Novus Biologicals Supplier No. NB11061020
Add to Cart
Add to Cart

Rat Monoclonal Antibody

PDGFR alpha Monoclonal specifically detects PDGFR alpha in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PDGFR alpha
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0004-3G28
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:400-1:800, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. P26618
Gene Alias alpha-type platelet-derived growth factor receptor, CD140 antigen-like family member A, CD140a, CD140a antigen, EC 2.7.10, EC 2.7.10.1, MGC74795, PDGFR2, PDGFRA/BCR fusion, PDGF-R-alpha, platelet-derived growth factor receptor, alpha polypeptide, rearranged-in-hypereosinophilia-platelet derived growth factor receptor alphafusion protein, RHEPDGFRA
Gene Symbols PDGFRA
Host Species Rat
Immunogen Mouse PDGFR alpha (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5156
Test Specificity This antibody recognizes mouse PDGF receptor alpha in western blot. No cross reactivity to mouse PDGF receptor beta.
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.