Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDGFRL Antibody (1008804) [DyLight 594], Novus Biologicals™
SDP

Catalog No. NB399054 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399054 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399054 Supplier Novus Biologicals Supplier No. FAB1723M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDGFRL Monoclonal antibody specifically detects PDGFRL in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen PDGFRL
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 1008804
Conjugate DyLight 594
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias PDGFR-like protein, PDGRL, platelet-derived growth factor receptor-like, platelet-derived growth factor receptor-like protein, platelet-derived growth factor-beta-like tumor suppressor, PRLTSPDGF receptor beta-like tumor suppressor
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human PDGFRL, Gln22-Ser375, Accession # Q15198
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5157
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.