Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pdi1p Antibody (38H8), Novus Biologicals™
SDP

Catalog No. NB018369 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018369 0.125 mL
NBP105391 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018369 Supplier Novus Biologicals Supplier No. NBP1053910.125ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pdi1p Monoclonal specifically detects Pdi1p in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Pdi1p
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Monoclonal
Clone 38H8
Concentration 6.6mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:500-1:1000
Formulation PBS with 10mM Sodium Azide
Gene Alias MFP 1, MFP1, PDI, PDI 1, PDI1, Protein disulfide isomerase, Thioredoxin related glycoprotein 1, TRG 1, TRG1
Host Species Mouse
Immunogen Yeast nuclear preparations
Molecular Weight of Antigen 58.2 kDa
Purification Method Tissue culture supernatant
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.