Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDK-1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP24413205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24413205 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24413205 Supplier Novus Biologicals Supplier No. NBP2441320.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDK-1 Polyclonal specifically detects PDK-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen PDK-1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:100000, Immunohistochemistry 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Gene Alias 3-phosphoinositide-dependent protein kinase 1, 3-phosphoinositide-dependent protein kinase 2 pseudogene, PDK1, PDPK2, PDPK2P, PkB kinase like gene 1
Gene Symbols PDPK1
Host Species Rabbit
Immunogen This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminal of human PDPK1 protein.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 5170
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.