Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDK4 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP107047A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP107047A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP107047A30 Supplier Novus Biologicals Supplier No. NBP107047AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDK4 Polyclonal specifically detects PDK4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDK4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial
Gene Symbols PDK4
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-ProtNo. Q16654]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5166
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.