Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDK4 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP107047G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP107047G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP107047G Supplier Novus Biologicals Supplier No. NBP107047G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDK4 Polyclonal specifically detects PDK4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDK4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q16654
Gene Alias [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial
Gene Symbols PDK4
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-Prot# Q16654]
Molecular Weight of Antigen 49 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Mitochondrial Markers, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5166
Test Specificity Human and Mouse. Predicted to react with bovine based on 100% sequence homology. Immunogen sequence has 94% homology to rat.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.