Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDLIM1 Mouse anti-Human, Clone: CPTC-PDLIM1-1, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP30751820UG

 View more versions of this product

Catalog No. NB122510


Only null left
Add to Cart

Description

Description

PDLIM1 Monoclonal specifically detects PDLIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

PDLIM1
Monoclonal
0.2 mg/ml
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
CLIM1carboxyl terminal LIM domain protein 1, CLP-36, CLP36carboxy terminal LIM domain protein 1, C-terminal LIM domain protein 1, elfin, enigma, hCLIM1, LIM domain protein CLP-36, PDZ and LIM domain 1, PDZ and LIM domain 1 (elfin), PDZ and LIM domain protein 1
Recombinant human full-length PDLIM1 protein
20 μg
Primary
Human
Purified
Western Blot, Immunohistochemistry (Paraffin)
CPTC-PDLIM1-1
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
9124
Store at 4°C.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.