Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDLIM1 Mouse anti-Human, Clone: CPTC-PDLIM1-1, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
| Antigen | PDLIM1 |
|---|---|
| Clone | CPTC-PDLIM1-1 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
PDLIM1 Monoclonal specifically detects PDLIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| PDLIM1 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| CLIM1carboxyl terminal LIM domain protein 1, CLP-36, CLP36carboxy terminal LIM domain protein 1, C-terminal LIM domain protein 1, elfin, enigma, hCLIM1, LIM domain protein CLP-36, PDZ and LIM domain 1, PDZ and LIM domain 1 (elfin), PDZ and LIM domain protein 1 | |
| Recombinant human full-length PDLIM1 protein | |
| Primary | |
| Store at 4°C. |
| CPTC-PDLIM1-1 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 9124 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title