Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDLIM2 Antibody (OTI11A12), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N273311P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273311P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273311P594 Supplier Novus Biologicals Supplier No. NBP273311PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDLIM2 Monoclonal specifically detects PDLIM2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen PDLIM2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI11A12
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ34715, MYSTIQUE, PDZ and LIM domain 2 (mystique), PDZ and LIM domain protein 2, PDZ-LIM protein mystique, SLIM
Gene Symbols PDLIM2
Host Species Mouse
Immunogen Full length recombinant protein of human PDLIM2 (NP_067643) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 64236.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.