Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDLIM4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219759 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219759 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219759 Supplier Novus Biologicals Supplier No. NBP219759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDLIM4 Polyclonal specifically detects PDLIM4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PDLIM4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. P50479
Gene Alias LIM domain protein, LIM protein RIL, PDZ and LIM domain 4, PDZ and LIM domain protein 4, Reversion-induced LIM protein, RILenigma homolog
Gene Symbols PDLIM4
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PDLIM4. The exact sequence is proprietary.
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8572
Test Specificity Human. Species reactivity based on sequence identity: Mouse (100%), Chicken (100%), Rabbit (100%), Xenopus laevis (100%), Bovine (100%), Xenopus tropicalis (100%), Zebrafish (93%), Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.