Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDLIM5 Antibody (OTI1H10), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP273317FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273317FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273317FR Supplier Novus Biologicals Supplier No. NBP273317FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDLIM5 Monoclonal specifically detects PDLIM5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen PDLIM5
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1H10
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Enh, ENH1, ENHLIM, Enigma homolog, enigma-like LIM domain protein, Enigma-like PDZ and LIM domains protein, PDZ and LIM domain 5, PDZ and LIM domain protein 5
Gene Symbols PDLIM5
Host Species Mouse
Immunogen Full length recombinant protein of human PDLIM5 (NP_006448) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10611.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.