Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDLIM7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106534 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106534 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106534 Supplier Novus Biologicals Supplier No. NBP106534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PDLIM7 Polyclonal specifically detects PDLIM7 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PDLIM7
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_005442.2
Gene Alias 1110003B01Rik, ENIGMAProtein enigma, LIM domain protein, LIM mineralization protein, LMP, LMP1, PDZ and LIM domain 7 (enigma), PDZ and LIM domain protein 7
Gene Symbols PDLIM7
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 407 and 457 of human LIM mineralization protein 1 using the numbering given in entry NP_005442.2 (GeneID 9260).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9260
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.