Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDRG1 Antibody (016), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB334027 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334027 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334027 Supplier Novus Biologicals Supplier No. NBP290269APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PDRG1 Monoclonal antibody specifically detects PDRG1 in Human samples. It is validated for ELISA

Specifications

Antigen PDRG1
Applications ELISA
Classification Monoclonal
Clone 016
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias C20orf126p53 and DNA damage regulated 1, chromosome 20 open reading frame 126, dJ310O13.3, p53 and DNA-damage regulated 1, PDRGp53 and DNA damage-regulated protein 1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human PDRG1 (Uniprot#: Q9NUG6, Met1-Gly133)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81572
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.