Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDSS2 Antibody (OTI1D12), Biotin, Novus Biologicals™
SDP

Catalog No. NBP273323B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273323B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273323B Supplier Novus Biologicals Supplier No. NBP273323B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDSS2 Monoclonal specifically detects PDSS2 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen PDSS2
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1D12
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias All-trans-decaprenyl-diphosphate synthase subunit 2, bA59I9.3, C6orf210decaprenyl-diphosphate synthase subunit 2, Candidate tumor suppressor protein, chromosome 6 open reading frame 210, Decaprenyl pyrophosphate synthase subunit 2, decaprenyl pyrophosphate synthetase subunit 2, DLP1hDLP1, EC 2.5.1.91, prenyl (decaprenyl) diphosphate synthase, subunit 2, subunit 2 of decaprenyl diphosphate synthase
Gene Symbols PDSS2
Host Species Mouse
Immunogen Full length recombinant protein of human PDSS2 (NP_065114) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 57107.0
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.