Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDX-1/IPF1 Antibody (OTI3H3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP273332A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273332A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273332A32 Supplier Novus Biologicals Supplier No. NBP273332AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDX-1/IPF1 Monoclonal specifically detects PDX-1/IPF1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen PDX-1/IPF1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3H3
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1
Gene Symbols PDX1
Host Species Mouse
Immunogen Full length recombinant protein of human PDX-1/IPF1 (NP_000200) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3651
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.