Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDX-1/IPF1 Antibody (PCRP-PDX1-2C11), PerCP, Novus Biologicals™
SDP

Catalog No. NB384050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384050 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384050 Supplier Novus Biologicals Supplier No. NBP308853PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDX-1/IPF1 Monoclonal antibody specifically detects PDX-1/IPF1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen PDX-1/IPF1
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-PDX1-2C11
Conjugate PerCP
Formulation PBS
Gene Alias Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1
Host Species Mouse
Immunogen Recombinant full-length human PDX-1/IPF1 protein (Uniprot: P52945)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3651
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.