Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDZD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546771
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA546771 100 μL
1 options

Catalog No. PIPA546771

Supplier: Invitrogen™ PA546771

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TRHIINTSSR LLNLRQVSKT RLSEPGTDLV EPSPKHTPNT SDNEGSDTEV.

PDZD8 plays a role in the regulation of cell morphology and cytoskeletal organization. It may inhibit herpes simplex virus 1 infection at an early stage and may promote retroviral infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDZD8
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PDZD8
Gene Accession No. Q8NEN9
Gene Alias A630041P07Rik; AV329468; AW228944; bA129M16.2; FLJ34427; PDZ domain containing 8; PDZ domain-containing protein 8; Pdzd8; PDZK8; Sarcoma antigen NY-SAR-84/NY-SAR-104
Gene Symbols PDZD8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PDZD8 (aa 935-984).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118987
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.