Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDZK1 Antibody, Novus Biologicals™
SDP

Catalog No. NB400149SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400149SS 0.025 mL
NB400149 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400149SS Supplier Novus Biologicals Supplier No. NB400149SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

PDZK1 Polyclonal specifically detects PDZK1 in Human, Mouse, Bovine, Rat (Negative) samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PDZK1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:250, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400
Formulation Whole antisera with 0.05% Sodium Azide
Gene Accession No. Q5T2W1
Gene Alias CAP70, CFTR-associated protein of 70 kDa, CLAMP, Na(+)/H(+) exchanger regulatory factor 3, Na/Pi cotransporter C-terminal-associated protein 1, naPi-Cap1, NHERF-3, NHERF3PDZD1Na(+)/H(+) exchange regulatory cofactor NHE-RF3, PDZ domain containing 1, PDZ domain-containing protein 1, PDZ-containing kidney protein 1, Sodium-hydrogen exchanger regulatory factor 3
Gene Symbols PDZK1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminus of human PDZK1. [UniProt# Q13113]
Molecular Weight of Antigen 57 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Diabetes Research, Endothelial Cell Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5174
Test Specificity Human, Bovine and mouse. Does not react with rat.
Target Species Human, Mouse, Bovine, Rat (Negative)
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.