Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDZK1 Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB102064 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102064 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102064 Supplier Novus Biologicals Supplier No. NBP297448AF594

Rabbit Polyclonal Antibody

PDZK1 Polyclonal specifically detects PDZK1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PDZK1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias CAP70, CFTR-associated protein of 70 kDa, CLAMP, Na(+)/H(+) exchanger regulatory factor 3, Na/Pi cotransporter C-terminal-associated protein 1, naPi-Cap1, NHERF-3, NHERF3PDZD1Na(+)/H(+) exchange regulatory cofactor NHE-RF3, PDZ domain containing 1, PDZ domain-containing protein 1, PDZ-containing kidney protein 1, Sodium-hydrogen exchanger regulatory factor 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PDZK1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Diabetes Research, Endothelial Cell Markers, Growth and Development, Lipid and Metabolism, Neuronal Cell Markers, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5174
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.